EMAIL THIS PAGE TO A FRIEND
Experimental eye research
Neuroprotective effects of C3 exoenzyme in excitotoxic retinopathy.
PMID 24928315
Abstract
The purpose of this study is to evaluate the neuroprotective effects of C3 exoenzyme (C3) on N-methyl-d-aspartate (NMDA)-induced retinopathy in rats. C3 was expressed in Escherichia. coli and purified by affinity chromatography. Immunofluorescence was performed in NIH 3T3 cells treated with C3 to verify the cellular uptake of the protein. NMDA was injected intravitreally into rat eyes with or without C3. At various time points after injection, eyes were enucleated. Hematoxylin/eosin staining was performed on retina cross-sections for morphological analysis. Survival and apoptosis of cells in the ganglion cell layer (GCL) were assessed by cresyl violet staining and terminal deoxynucleotidyl transferase dUTP nick end labeling (TUNEL) on retina flat-mounts. RhoA levels in retina cells were evaluated by Western blot to detect C3 uptake in vivo. The cellular uptake of C3 was verified by immunofluorescence. Damage including a decrease in inner plexiform layer (IPL) thickness and reduction of cell density in the GCL, corresponding to apoptosis of neurons, was induced by intravitreal injection of NMDA. Protection against this damage was observed following co-injection of C3 and NMDA. RhoA ADP-ribosylation induced by C3 was confirmed by Western blot. Our results suggest that C3 exerts neuroprotective effects against excitotoxic damage induced by NMDA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3700853 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700863 Anti-Rabbit IgG (Fc specific)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700852 Anti-Rabbit IgG (Fc specific)-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700949 Anti-Rabbit IgG (H+L), F(ab′)2 fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700978 Anti-Rabbit IgG (H+L), F(ab) fragment-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700972 Anti-Rabbit IgG (H+L), F(ab) fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700941 Anti-Rabbit IgG (H+L)-Peroxidase antibody produced in chicken, affinity isolated antibody, lyophilized powder
SAB3700934 Anti-Rabbit IgG (H+L)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700920 Anti-Rabbit IgG (H+L)-Peroxidase antibody produced in sheep, affinity isolated antibody, lyophilized powder
SAB3700842
Anti-Rabbit IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700832 Anti-Rabbit IgG F(ab′)2, highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700831 Anti-Rabbit IgG F(ab′)2-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
T2580000
Tropicamide, European Pharmacopoeia (EP) Reference Standard
C17H20N2O2