EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Non-viral reprogramming of fibroblasts into induced pluripotent stem cells by Sleeping Beauty and piggyBac transposons.
PMID 24928388
Abstract
The generation of induced pluripotent stem (iPS) cells represents a promising approach for innovative cell therapies. The original method requires viral transduction of several reprogramming factors, which may be associated with an increased risk of tumorigenicity. Transposition of reprogramming cassettes represents a recent alternative to viral approaches. Since binary transposons can be produced as common plasmids they provide a safe and cost-efficient alternative to viral delivery methods. Here, we compared the efficiency of two different transposon systems, Sleeping Beauty (SB) and piggyBac (PB), for the generation of murine iPS. Murine fibroblasts derived from an inbred BL/6 mouse line carrying a pluripotency reporter, Oct4-EGFP, and fibroblasts derived from outbred NMRI mice were employed for reprogramming. Both transposon systems resulted in the successful isolation of murine iPS cell lines. The reduction of the core reprogramming factors to omit the proto-oncogene c-Myc was compatible with iPS cell line derivation, albeit with reduced reprogramming efficiencies. The transposon-derived iPS cells featured typical hallmarks of pluripotency, including teratoma growth in immunodeficient mice. Thus SB and PB transposons represent a promising non-viral approach for iPS cell derivation.