EMAIL THIS PAGE TO A FRIEND
European Journal of Pharmaceutics and Biopharmaceutics
Tablets of pre-liposomes govern in situ formation of liposomes: concept and potential of the novel drug delivery system.
PMID 24929211