EMAIL THIS PAGE TO A FRIEND
Acta Pharmacologica Sinica
A cell-based, high-throughput homogeneous time-resolved fluorescence assay for the screening of potential ?-opioid receptor agonists.
PMID 24930486