EMAIL THIS PAGE TO A FRIEND
Oncotarget
Dual Aurora A and JAK2 kinase blockade effectively suppresses malignant transformation.
PMID 24930769
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU028351 MISSION® esiRNA, esiRNA targeting mouse Aurka (esiRNA1)