EMAIL THIS PAGE TO A FRIEND
The Journal of pathology
Odd-skipped related 1 is a novel tumour suppressor gene and a potential prognostic biomarker in gastric cancer.
PMID 24931004
Abstract
We report that the odd-skipped related 1 (OSR1) gene encoding a zinc-finger transcription factor was preferentially methylated in gastric cancer by genome-wide methylation screening. OSR1 expression was frequently silenced or down-regulated in gastric cancer cell lines. OSR1 expression was also significantly down-regulated at both mRNA and protein levels in primary gastric cancer tissues compared with adjacent normal tissues. The silencing or down-regulation of OSR1 was closely associated with promoter hypermethylation. Overexpression of OSR1 significantly inhibited cell growth, arrested the cell cycle, and induced apoptosis in the gastric cancer cell lines AGS, MKN28, and MGC803. Conversely, knockdown of OSR1 by OSR1-short hairpin RNA significantly enhanced cell growth, promoted the cell cycle, and inhibited apoptosis in the normal gastric epithelial cell line GES1. The dual-luciferase reporter assay revealed that OSR1 activated p53 transcription and repressed the T-cell factor (TCF)/lymphoid enhancer factor (LEF). Complementary DNA expression array and western blotting showed that OSR1 increased the expression of nuclear p53, p21, Fas, and death receptor-5, and suppressed the expression of cyclin D1 and cyclin-dependent kinase 4 in the p53 signalling pathway. In addition, OSR1 suppressed the expression of cytoplasmic β-catenin, TCF-1, and LEF1 in the Wnt/β-catenin signalling pathway. OSR1 methylation was detected in 51.8% of primary gastric cancer patients (85 of 164) by bisulphite genomic sequencing. Multivariate Cox regression analysis showed that OSR1 methylation was an independent predictor of poor survival. Kaplan-Meier survival curves revealed that OSR1 methylation was associated with shortened survival in TNM stage I-III patients. In conclusion, OSR1 acts as a functional tumour suppressor through the transcriptional activation of p53 and repression of TCF/LEF in gastric cancer. Detection of OSR1 methylation may serve as a potential biomarker of the early stage of gastric cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4503743
Anti-OSR1 antibody produced in rabbit, affinity isolated antibody
SAB4503744
Anti-OXSR1 antibody produced in rabbit, affinity isolated antibody
HPA008237
Anti-OXSR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411706
Anti-OXSR1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1408873 Monoclonal Anti-OSR1 antibody produced in mouse, clone 2B4, ascites fluid, solution
SAB1408875 Monoclonal Anti-OSR1 antibody produced in mouse, clone 3H8, purified immunoglobulin, buffered aqueous solution
SAB1408876
Monoclonal Anti-OSR1 antibody produced in mouse, clone 1G9, purified immunoglobulin, buffered aqueous solution
SAB1408878 Monoclonal Anti-OSR1 antibody produced in mouse, clone 1C6, purified immunoglobulin, buffered aqueous solution
SAB1405355
Monoclonal Anti-OSR1, (N-terminal) antibody produced in mouse, clone 1D5, purified immunoglobulin, buffered aqueous solution
WH0009943M16
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 3A8, purified immunoglobulin, buffered aqueous solution
SAB1404767
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 1B9, purified immunoglobulin, buffered aqueous solution
SAB1410032
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 4G9, purified immunoglobulin, buffered aqueous solution
SAB1410033
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 5E1, purified immunoglobulin, buffered aqueous solution
SAB1410034
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 2E9, purified immunoglobulin, buffered aqueous solution
SAB1410035
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 1F6, purified immunoglobulin, buffered aqueous solution
SAB1410036
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 4D12, purified immunoglobulin, buffered aqueous solution
SAB1410037
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 2A2-1A2, purified immunoglobulin, buffered aqueous solution
SAB1410038
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 4C12, purified immunoglobulin, buffered aqueous solution
SAB1410039
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 1C8, purified immunoglobulin, buffered aqueous solution
SAB1410041
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 5D5, purified immunoglobulin, buffered aqueous solution
SAB1410042
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 3F6, purified immunoglobulin, buffered aqueous solution
SAB1410043
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 2A5, purified immunoglobulin, buffered aqueous solution
SAB1410044
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 1B10, purified immunoglobulin, buffered aqueous solution
SAB1410045
Monoclonal Anti-OXSR1 antibody produced in mouse, clone 1D10, purified immunoglobulin, buffered aqueous solution
SAB2106818
Anti-OXSR1 antibody produced in rabbit, affinity isolated antibody