EMAIL THIS PAGE TO A FRIEND
Journal of Clinical Research in Pediatric Endocrinology
Generation of monocyte-derived insulin-producing cells from non-human primates according to an optimized protocol for the generation of PCMO-derived insulin-producing cells.
PMID 24932602