EMAIL THIS PAGE TO A FRIEND
Fitoterapia
Lignan formation in hairy root cultures of Edelweiss (Leontopodium nivale ssp. alpinum (Cass.) Greuter).
PMID 24932777
Abstract
A hairy root line of Edelweiss (Leontopodium nivale ssp. alpinum (Cass.) Greuter) was obtained upon transformation with Agrobacterium rhizogenes strain ATCC15834. Elicitation of this line with silver nitrate, sucrose, methyl jasmonate and yeast extract at various concentrations in most cases resulted in a stimulation of lignan biosynthesis. Through elicitation with 6% sucrose the roots accumulated the pharmacologically active lignans leoligin and 5-methoxy-leoligin at levels of 0.0678% and 0.0372%, respectively, without significant growth inhibition. These lignan levels were comparable to those found in intact roots of cultivated Edelweiss. The biotechnological production of leoligin could be an attractive option for the continuous, field culture-independent production of the valuable secondary metabolites leoligin and 5-methoxy-leoligin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
392707
Methyl jasmonate, 95%
C13H20O3
W341002
Methyl jasmonate, ≥98%, stabilized, FG
C13H20O3