EMAIL THIS PAGE TO A FRIEND
Journal of Ethnopharmacology
The mechanism of hepatoprotective effect of sesquiterpene rich fraction from Cichorum glandulosum Boiss. et Huet on immune reaction-induced liver injury in mice.
PMID 24933227