EMAIL THIS PAGE TO A FRIEND
The New York state dental journal
Effect of irrigation with Tetraclean on bacterial leakage of obturated root canals.
PMID 24933773
Abstract
The purpose of the study presented here was to evaluate the effect of Tetraclean, Hypoclean, Chlor-XTRA, 2% chlorhexidine and 6% sodium hypochlorite/17% EDTA as a final irrigant on bacterial leakage of the root canal. One hundred and fifty-five extracted human maxillary central incisors were randomly divided into five experimental groups of 25 teeth each and two control groups of 15 teeth each. The root canals in each group were irrigated with 2 ml of the relevant irrigant between each filing. The root canals in group 5 were irrigated with 5 ml of 17% EDTA at the end of root canal preparation. The teeth in each group were obturated with gutta-percha and AH-26 sealer. Positive control teeth were obturated with a single gutta-percha cone without sealer, and negative controls were obturated in the same way as experimental groups. The coronal portion of each root was placed in contact with inoculum of Enterococcus faecalis in Brain Heart Infusion (BHI) culture media. Findings showed that the mean number of days for bacterial penetration in the Tetraclean group was greater than for other experimental groups. On the other hand, the Chlor-XTRA Vista group showed the fewest mean number of days for bacterial leakage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
452386
Bismuth, beads, 1-5 mm, 99.999% trace metals basis
Bi
95372
Bismuth, granular, ≥99.99% trace metals basis
Bi
556130
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Bi
264008
Bismuth, powder, −100 mesh, ≥99.99% trace metals basis
Bi
265462
Bismuth, powder, −100 mesh, 99% trace metals basis
Bi
265470
Bismuth, shot, 4-30 mesh, 99.9% trace metals basis
Bi
GF86561354
Bismuth, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF85045631
Bismuth, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF80139438
Bismuth, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF81875447
Bismuth, rod, 120mm, diameter 10.0mm, cast sticks, 100%
Bi
GF71052432
Bismuth, rod, 140mm, diameter 10.0mm, cast sticks, 100%
Bi
GF60966366
Bismuth, rod, 70mm, diameter 10.0mm, cast sticks, 100%
Bi
GF89918500
Bismuth, foil, 25x25mm, thickness 0.25mm, 100%
Bi
GF71570550
Bismuth, foil, 25x25mm, thickness 0.5mm, 100%
Bi
GF58555581
Bismuth, foil, 25x25mm, thickness 1.0mm, 100%
Bi
GF52383506
Bismuth, foil, 25x25mm, thickness 1.0mm, 99.97%
Bi
GF31001960
Bismuth, foil, 25x25mm, thickness 2.0mm, 100%
Bi
GF39813419
Bismuth, foil, 50x50mm, thickness 0.25mm, 100%
Bi
GF46319340
Bismuth, foil, 50x50mm, thickness 0.5mm, 100%
Bi
GF19448017
Bismuth, foil, 50x50mm, thickness 1.0mm, 100%
Bi
GF97656238
Bismuth, foil, 50x50mm, thickness 1.0mm, 99.97%
Bi
GF75246183
Bismuth, foil, 50x50mm, thickness 2.0mm, 100%
Bi
GF46489135
Bismuth, foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.97%
Bi
GF86892441
Bismuth, foil, light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF40931546
Bismuth, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF35246831
Bismuth, foil, light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.97%
Bi
GF95571995
Bismuth, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF12550935
Bismuth, foil, light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 99.97%
Bi
GF09655263
Bismuth, foil, light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF71673060
Bismuth, foil, light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi