EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Weight loss interventions and breast cancer survival: the time is now.
PMID 24934785