EMAIL THIS PAGE TO A FRIEND
Tissue engineering. Part A
Enhancement of cell ingrowth, proliferation, and early differentiation in a three-dimensional silicon carbide scaffold using low-intensity pulsed ultrasound.
PMID 24935158
Abstract
Concerns over the use of autografts or allografts have necessitated the development of biomaterials for bone regeneration. Various studies have been performed to optimize the cultivation of osteogenic cells using osteoconductive porous scaffolds. The aim of this study was to evaluate the osteogenic efficiency of bone cell ingrowth, proliferation, and early differentiation in a silicon carbide (SiC) porous ceramic scaffold promoted with low-intensity pulsed ultrasound. MC3T3-E1 mouse preosteoblasts were seeded onto scaffolds and cultured for 4 and 7 days with daily of 20-min ultrasound treatment. The cells were evaluated for cell attachment, morphology, viability, ingrowth depth, volumetric proliferation, and early differentiation. After 4 and 7 days of culture and ultrasound exposure, the cell density was higher in the ultrasound-treated group compared with the sham-treated group on SiC scaffolds. The cell ingrowth depths inside the SiC scaffolds were 149.2±27.3 μm at 1 day, 310.1±12.6 μm for the ultrasound-treated group and 248.0±19.7 μm for the sham control at 4 days, and 359.6±18.5 μm for the ultrasound-treated group and 280.0±17.7 μm for the sham control at 7 days. They were significantly increased, that is, 25% (p=0.0029) and 28% (p=0.0008) increase, respectively, with ultrasound radiation force as compared with those in sham control at 4 and 7 days postseeding. The dsDNA contents were 583.5±19.1 ng/scaffold at 1 day, 2749.9±99.9 ng/scaffold for the ultrasound-treated group and 2514.9±114.7 ng/scaffold for the sham control at 4 days, and 3582.3±325.3 ng/scaffold for the ultrasound-treated group and 2825.7±134.3 ng/scaffold for the sham control at 7 days. There was a significant difference in the dsDNA content between the ultrasound- and sham-treated groups at 4 and 7 days. The ultrasound-treated group with the SiC construct showed a 9% (p=0.00029) and 27% (p=0.00017) increase in the average dsDNA content at 4 and 7 days over the sham control group, respectively. Alkaline phosphatase activity was significantly increased by the treatment of ultrasound at 4 (p=0.012) and 7 days (p=0.035). These results suggested that ultrasound treatment with low-intensity acoustic energy facilitated the cellular ingrowth and enhanced the proliferation and early differentiation of osteoblasts in SiC scaffolds.