EMAIL THIS PAGE TO A FRIEND
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons
Pretransplant IgG reactivity to apoptotic cells correlates with late kidney allograft loss.
PMID 24935695
Abstract
Preexisting serum antibodies have long been associated with graft loss in transplant recipients. While most studies have focused on HLA-specific antibodies, the contribution of non-HLA-reactive antibodies has been largely overlooked. We have recently characterized mAbs secreted by B cell clones derived from kidney allograft recipients with rejection that bind to apoptotic cells. Here, we assessed the presence of such antibodies in pretransplant serum from 300 kidney transplant recipients and examined their contribution to the graft outcomes. Kaplan-Meier survival analysis revealed that patients with high pretransplant IgG reactivity to apoptotic cells had a significantly increased rate of late graft loss. The effect was only apparent after approximately 1 year posttransplant. Moreover, the association between pretransplant IgG reactivity to apoptotic cells and graft loss was still significant after excluding patients with high reactivity to HLA. This reactivity was almost exclusively mediated by IgG1 and IgG3 with complement fixing and activating properties. Overall, our findings support the view that IgG reactive to apoptotic cells contribute to presensitization. Taking these antibodies into consideration alongside anti-HLA antibodies during candidate evaluation would likely improve the transplant risk assessment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3773 Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal, clone HP-6017, purified from hybridoma cell culture
RAB0001
Human IgG ELISA Kit, for serum, plasma
G2904 Monoclonal Anti-Goat/Sheep IgG antibody produced in mouse, clone GT-34, ascites fluid
A8062 Monoclonal Anti-Goat/Sheep IgG–Alkaline Phosphatase antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous glycerol solution
B3148 Monoclonal Anti-Goat/Sheep IgG–Biotin antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
F4891 Monoclonal Anti-Goat/Sheep IgG−FITC antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
A9452 Monoclonal Anti-Goat/Sheep IgG–Peroxidase antibody produced in mouse, clone GT-34, purified immunoglobulin, lyophilized powder
I6135 Monoclonal Anti-Human IgG (Fab specific) antibody produced in mouse, clone GG-6, ascites fluid
I6260 Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone GG-7, ascites fluid
I6760 Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone HP-6017, ascites fluid
F5016 Monoclonal Anti-Human IgG (Fc specific)−FITC antibody produced in mouse, clone HP-6017, purified immunoglobulin, buffered aqueous solution
I5885 Monoclonal Anti-Human IgG (γ-chain specific) antibody produced in mouse, clone GG-5, ascites fluid
I9513 Monoclonal Anti-Human IgG2 (Fc specific) antibody produced in mouse, clone HP-6002, ascites fluid
I9888 Monoclonal Anti-Human IgG4 (pFc′ specific) antibody produced in mouse, clone HP-6023, ascites fluid
R0761 Monoclonal Anti-Rat IgG2a antibody produced in mouse, clone R2A-2, ascites fluid
RAB0803 Mouse IgG2a ELISA Kit
RAB0805 Mouse IgG3 ELISA Kit