EMAIL THIS PAGE TO A FRIEND
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology
Histomorphometry in antigen-induced arthritis of the rabbit temporomandibular joint.
PMID 24935724
Abstract
Juvenile idiopathic arthritis (JIA) of the temporomandibular joint (TMJ) can cause severe growth disturbances of the craniomandibular system. Antigen-induced arthritis (AIA) of the rabbit TMJ is simulating the inflammatory process of the TMJ in JIA. The aim of this study was to investigate the effect of a systemic administration of the tumor necrosis factor-alpha (TNF-α) antagonist etanercept on AIA in rabbits by means of three different histological staining methods. After sensitization, a bilateral arthritis of the TMJ was induced and maintained by repeated intra-articular administrations of ovalbumin in 12 New Zealand white rabbits aged 10 weeks. From the 13th week of age, 6 of the 12 rabbits received weekly subcutaneous injections of etanercept, and the other 6 animals remained without therapy. Another 6 animals served as controls, receiving no treatment or intra-articular injections at all. After euthanasia at the age of 22 weeks, all TMJs were retrieved en bloc. Sagittal sections were cut and stained with hematoxylin-eosin (H-E), Safranin-O for the evaluation of the Mankin score, and tartrate-resistant acid phosphatase (TRAP). In the arthritis group, a chronic inflammation with degeneration of the articular cartilage was visible. In the etanercept group, the signs of cartilage degeneration were significantly reduced but present. In contrast, the joints in the control group were inconspicuous. A strong correlation between the Mankin score and TRAP-positive cells could be found. Antigen-induced arthritis causes severe damage in the TMJ of young rabbits. An improvement seems to be achievable by a systemic administration of etanercept.