EMAIL THIS PAGE TO A FRIEND
Prostaglandins, leukotrienes, and essential fatty acids
The effect of dietary omega-3 polyunsaturated fatty acids on plasma lipids and lipoproteins of C57BL/6 mice is age and sex specific.
PMID 24935799
Abstract
There is clear evidence of the effects of sex and age on the prevalence of cardiovascular disease. We investigated the interactions of dietary omega (n)-3 polyunsaturated fatty acids (PUFA), sex, and age on plasma lipids and lipoproteins in the offspring of C57BL/6 mice exposed to high, medium, or low n-3 PUFA at weaning and 16 weeks postweaning. There was an increase in plasma triglycerides from weaning to 16 weeks in male and female offspring; however, the high n-3 PUFA group showed a reduction in triglycerides in both sexes at 16 weeks. High n-3 PUFA caused an increase in plasma LDL-cholesterol from weaning to 16 weeks in male offspring; however, the LDL particle size was significantly larger in the high n-3 PUFA group. Plasma from male mice showed higher cholesterol efflux compared to females; high n-3 PUFA increased cholesterol efflux. Thus the effects of n-3 PUFA are age and sex dependent.