EMAIL THIS PAGE TO A FRIEND
Planta medica
Cytotoxic coumarins from Toddalia asiatica.
PMID 24936959
Abstract
Three new coumarins and 13 known compounds were isolated from the stem bark of Toddalia asiatica. Compounds 1, 3, 8, and 9 showed cytotoxicity against the NCI-H187 cell line with IC50 values ranging from 6 to 9 µg/mL. Compounds 4 and 9 exhibited cytotoxicity against the MCF-7 cell line with IC50 values of 3.17 and 9.79 µg/mL, respectively. Compound 9 also displayed cytotoxic activity against KB cells with an IC50 value of 8.63 µg/mL. In addition, compound 14 showed antimalarial activity against Plasmodium falciparum with an IC50 value of 3.66 µg/mL. Compounds 5, 9, and 16 exhibited antituberculosis activity against Mycobacterium tuberculosis with MIC values of 50, 50, and 25 µg/mL, respectively.