EMAIL THIS PAGE TO A FRIEND
Translational Psychiatry
Reversal of autism-like behaviors and metabolism in adult mice with single-dose antipurinergic therapy.
PMID 24937094
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09859
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
H5NO