EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Decarboxylative polymerization of 2,6-naphthalenedicarboxylic acid at surfaces.
PMID 24937642
Abstract
Metal-catalyzed polymerization of 2,6-naphthalenedicarboxylic acid (NDCA) to form poly-2,6-naphthalenes at various surfaces is reported. Polymerizations occur via initial formal dehydrogenation of self-assembled diacids with subsequent decarboxylation to give polymeric bisnaphthyl-Cu species at elevated temperature as intermediate structures (<160 °C). Further temperature increase eventually leads to poly-naphthalenes via reductive elimination. It is demonstrated that the Cu(111) surface works most efficiently to conduct such polymerizations as compared to the Au(111), Ag(111), Cu(100), and Cu(110) surfaces. Poly-2,6-naphthalene with a chain length of over 50 nm is obtained by using this approach. The decarboxylative coupling of aromatic diacids is a very promising tool which further enlarges the portfolio of reactions allowing for on-surface polymerizations and novel organometallic systems preparations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
301353
2,6-Naphthalenedicarboxylic acid, 95%
C12H8O4