PloS one
Antioxidant capacities, phenolic profile and cytotoxic effects of saxicolous lichens from trans-Himalayan cold desert of Ladakh.
PMID 24937759
Abstract
Fourteen saxicolous lichens from trans-Himalayan Ladakh region were identified by morpho-anatomical and chemical characteristics. The n-hexane, methanol and water extracts of the lichens were evaluated for their antioxidant capacities. The lichen extracts showing high antioxidant capacities and rich phenolic content were further investigated to determine their cytotoxic activity on human HepG2 and RKO carcinoma cell lines. The ferric reducing antioxidant power (FRAP), 2,2'-azinobis-(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt (ABTS), 1,1-diphenyl-2-picrylhydrazyl (DPPH) and nitric oxide (NO) radical scavenging capacities and β-carotene-linoleic acid bleaching property exhibited analogous results where the lichen extracts showed high antioxidant action. The lichen extracts were also found to possess good amount of total proanthocyanidin, flavonoid and polyphenol. The methanolic extract of Lobothallia alphoplaca exhibited highest FRAP value. Methanolic extract of Xanthoparmelia stenophylla showed the highest ABTS radical scavenging capacity. The n-hexane extract of Rhizoplaca chrysoleuca exhibited highest DPPH radical scavenging capacity. Highest antioxidant capacity in terms of β-carotene linoleic acid bleaching property was observed in the water extract of Xanthoria elegans. Similarly, Melanelia disjuncta water extract showed highest NO scavenging capacity. Among n-hexane, methanol and water extracts of all lichens, the methanolic extract of Xanthoparmelia mexicana showed highest total proanthocyanidin, flavonoid and polyphenol content. From cytotoxic assay, it was observed that the methanolic extracts of L. alphoplaca and M. disjuncta were exhibiting high cytotoxic effects against cancer cell growth. Similarly, the water extract of Dermatocarpon vellereum, Umbilicaria vellea, X. elegans and M. disjuncta and the methanolic extract of M. disjuncta and X. stenophylla were found to possess high antioxidant capacities and were non-toxic and may be used as natural antioxidants for stress related problems. Our studies go on to prove that the unique trans-Himalayan lichens are a hitherto untapped bioresource with immense potential for discovery of new chemical entities, and this biodiversity needs to be tapped sustainably.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
296309
1,4-Dioxane, anhydrous, 99.8%
C4H8O2
533971
1,4-Dioxane, histological grade, ≥99%
C4H8O2
76887
1,4-Dioxane, analytical standard
C4H8O2
07-5360
1,4-Dioxane, SAJ first grade
C4H8O2
07-5390
1,4-Dioxane, for HPLC
C4H8O2
PHR1549
1,4-Dioxane, Pharmaceutical Secondary Standard; Certified Reference Material
C4H8O2
17779
2,3,5-Triphenyl-tetrazolium chloride solution, for microbiology
C19H15ClN4
93285
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0%
C18H12N6
T1253
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. (of Fe), ≥98%
C18H12N6
34750
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
C15H24O
03762
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph.Eur.
C15H24O
03382
Aluminum atomic spectroscopy standard concentrate 10.00 g Al, 10.00 g/L, for 1 l standard solution, analytical standard
AlCl3
563919
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
AlCl3
294713
Aluminum chloride, 99.99% trace metals basis
AlCl3
449598
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
AlCl3
495352
Aluminum chloride, ReagentPlus®, anhydrous, ≥99.9% trace metals basis
AlCl3
06230
Aluminum chloride, purum, anhydrous, ≥98.0% (AT)
AlCl3
249882
Aluminum chloride solution, 1.0 M in nitrobenzene
AlCl3
06154
Aluminum standard for AAS, analytical standard, traceable to BAM
AlCl3
1082708
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
C15H24O
PHR1117
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
C15H24O
91215
Gallic acid, certified reference material, TraceCERT®
C7H6O5
451649
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Cl3Fe
701122
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Cl3Fe
361003
Iron(III) chloride on silica gel, powder, extent of labeling: 5 wt. % loading
Cl3Fe
710857
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Cl3Fe
77634
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
C8H8O3
P5592
Potassium peroxodisulfate, BioXtra, ≥99.0%
K2O8S2
60489
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
K2O8S2
379824
Potassium persulfate, 99.99% trace metals basis
K2O8S2