EMAIL THIS PAGE TO A FRIEND
International Journal of Oncology
Ginsenoside Rg3 inhibition of vasculogenic mimicry in pancreatic cancer through downregulation of VE?cadherin/EphA2/MMP9/MMP2 expression.
PMID 24938458