EMAIL THIS PAGE TO A FRIEND
Journal of neuroscience research
Enzyme replacement therapy attenuates disease progression in a canine model of late-infantile neuronal ceroid lipofuscinosis (CLN2 disease).
PMID 24938720
Abstract
Using a canine model of classical late-infantile neuronal ceroid lipofuscinosis (CLN2 disease), a study was conducted to evaluate the potential pharmacological activity of recombinant human tripeptidyl peptidase-1 (rhTPP1) enzyme replacement therapy administered directly to the cerebrospinal fluid (CSF). CLN2 disease is a hereditary neurodegenerative disorder resulting from mutations in CLN2, which encodes the soluble lysosomal enzyme tripeptidyl peptidase-1 (TPP1). Infants with mutations in both CLN2 alleles develop normally but in the late-infantile/early-childhood period undergo progressive neurological decline accompanied by pronounced brain atrophy. The disorder, a form of Batten disease, is uniformly fatal, with clinical signs starting between 2 and 4 years of age and death usually occurring by the early teenage years. Dachshunds homozygous for a null mutation in the canine ortholog of CLN2 (TPP1) exhibit a similar disorder that progresses to end stage at 10.5-11 months of age. Administration of rhTPP1 via infusion into the CSF every other week, starting at approximately 2.5 months of age, resulted in dose-dependent significant delays in disease progression, as measured by delayed onset of neurologic deficits, improved performance on a cognitive function test, reduced brain atrophy, and increased life span. Based on these findings, a clinical study evaluating the potential therapeutic value of rhTPP1 administration into the CSF of children with CLN2 disease has been initiated.