EMAIL THIS PAGE TO A FRIEND
Biometals: An International Journal on the Role of Metal Ions in Biology, Biochemistry, And Medicine
Streptococcus pneumoniae secretes a glyceraldehyde-3-phosphate dehydrogenase, which binds haemoglobin and haem.
PMID 24938797