EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Ultra-preconcentration and determination of selected pharmaceutical and personal care products in different water matrices by solid-phase extraction combined with dispersive liquid-liquid microextraction prior to ultra high pressure liquid chromatography tandem mass spectrometry analysis.
PMID 24939087
Abstract
Pharmaceutical and personal care products (PPCPs) are one of the most important classes of emerging contaminants. The potential of ecological and environmental impacts associated with PPCPs are of particular concern because they continually penetrate the aquatic environment. This work describes a novel ultra-preconcentration technique for the rapid and highly sensitive analysis of selected PPCPs in environmental water matrices at ppt levels. Selected PPCPs were rapidly extracted and concentrated from large volumes of aqueous solutions (500 and 250mL) by solid-phase extraction combined with dispersive liquid-liquid microextraction (SPE-DLLME) and then analyzed using UHPLC-MS/MS. Experimental parameters were carefully investigated and optimized to achieve the best SPE-DLLME efficiency and higher enrichment factors. The best results were obtained using the ternary mixture acetonitrile/methanol/dichloromethane 3:3:4, v/v/v, both as SPE eluent and DLLME extractant/dispersive mixture. DLLME aqueous solution (5% NaCl, 10mgL(-1) TBAB) was also modified to improve the extraction efficiency of more hydrophilic PPCPs. Under the optimal conditions, an exhaustive extraction for most of the investigated analytes (recoveries >70%), with a precision (RSD <10%) and very high enrichment factors were attained for different aqueous matrices (drinking, sea, river and wastewater). Method detection and quantification limits were at very low ppt levels and below 1 and 3ngL(-1), respectively, for 15 of selected PPCPs. The proposed analytical procedure offers numerous advantages such as the simplicity of operation, rapidity, a high enrichment factor and sensitivity. So it is suitable for monitoring and studies of occurrence of PPCPs in different environmental compartments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284785
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
C14H14O3
T16403
2,2′,4,4′-Tetrahydroxybenzophenone, 97%
C13H10O5
126217
2,4-Dihydroxybenzophenone, 99%
C13H10O3
415820
2-Ethylhexyl 2-cyano-3,3-diphenylacrylate, 97%
C24H27NO2
W514500
2-Ethylhexyl salicylate, ≥99%
C15H22O3
52184
2-Ethylhexyl salicylate, analytical standard
C15H22O3
437379
2-Ethylhexyl salicylate, 99%
C15H22O3
01479
2-Hydroxy-4-methoxybenzophenone, certified reference material, TraceCERT®
C14H12O3
H36206
2-Hydroxy-4-methoxybenzophenone, 98%
C14H12O3
105937
3,4,4′-Trichlorocarbanilide, 99%
C13H9Cl3N2O
PHR1303
3,4,4` Trichlorocarbanilide (Triclocarban), Pharmaceutical Secondary Standard; Certified Reference Material
C13H9Cl3N2O
H20202
4-Hydroxybenzophenone, 98%
C13H10O2
380709
Benzyl 4-hydroxybenzoate, 99%
C14H12O3
54680
Butyl 4-hydroxybenzoate, ≥99.0% (GC)
C11H14O3
W220302
Butyl 4-hydroxybenzoate, ≥99%
C11H14O3
B1217000 Butyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
C11H14O3
1084000 Butylparaben, United States Pharmacopeia (USP) Reference Standard
C11H14O3
PHR1022 Butylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
C11H14O3
C4024
Carbamazepine, powder
C15H12N2O
94496
Carbamazepine, analytical standard
C15H12N2O
C8981
Carbamazepine, meets USP testing specifications
C15H12N2O
C0450000
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
C15H12N2O
1093001
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
C15H12N2O
PHR1067
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
C15H12N2O
36542
DEET, PESTANAL®, analytical standard
C12H17NO
442541
DEET, analytical standard
C12H17NO
111988
Ethyl 4-hydroxybenzoate, ReagentPlus®, 99%
C9H10O3
E2425000
Ethyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
C9H10O3
PHR1011
Ethylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
C9H10O3
1267000
Ethylparaben, United States Pharmacopeia (USP) Reference Standard
C9H10O3