EMAIL THIS PAGE TO A FRIEND
International Journal of Pharmaceutics
Evaluation of synergistic effect of biodegradable polymeric nanoparticles and aluminum based adjuvant for improving vaccine efficacy.
PMID 24939616