EMAIL THIS PAGE TO A FRIEND
Pharmaceutical Research
A novel strategy for pharmaceutical cocrystal generation without knowledge of stoichiometric ratio: myricetin cocrystals and a ternary phase diagram.
PMID 24939640