EMAIL THIS PAGE TO A FRIEND
Journal of medicinal food
Anti-tumor effects of flavonoids from the ethnic medicine Docynia delavayi (Franch.) Schneid. and its possible mechanism.
PMID 24940817
Abstract
This study investigated the active components and the anti-tumor efficacy and mechanisms of the flavonoids from Docynia delavayi (Franch.) Schneid. (DDS). MTT assay was used to examine the growth inhibitory effects of the four flavonoids, including chrysin, quercetin, naringenin, and avicularin that were isolated from the rhizome of DDS, on human hematomas cell (HepG2), esophageal carcinoma cell (EC109), human cervical adenocarcinoma cell (Hela), human colon adenocarcinoma cell (SW480), and African green monkey kidney cell (Vero cells). The anti-tumor mechanism of chrysin on HepG2 was further investigated by the methods of fluorescence staining, flow cytometry, and immunoblotting. The results showed that the inhibitory activity of chrysin was much stronger than the other three flavonoids on HepG2, EC109, Hela, and SW480 cells for 48 h treatment in vitro. Moreover, no inhibiting effect of chrysin on the proliferation of normal cells (Vero cells) was observed. Further study revealed that chrysin caused HepG2 cell shrinkage, membrane blebbing, and apoptotic body formation, all of which were typical characteristics of apoptosis programmed cell death. Flow cytometric analysis demonstrated that chrysin increased the sub G0/G1 population, which indicated the increased cell apoptosis, thus preventing cells from entering the S phase as the population in G2/M or S phase declined; whereas in G0/G1 phase, it increased. In addition, immunoblot results showed that chrysin significantly increased the expression levels of caspase-3 and Bax proteins, and it decreased the expression level of B-cell lymphoma/leukemia-2 (Bcl-2) protein. These findings indicate that chrysin is the major flavonoid present in DDS, and it induces HepG2 cell death via apoptosis, probably through the participation of caspase-3, Bax, and Bcl-2 proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1033203
Ampicillin sodium, United States Pharmacopeia (USP) Reference Standard
C16H18N3NaO4S
PHR1424
Ampicillin Sodium, Pharmaceutical Secondary Standard; Certified Reference Material
C16H18N3NaO4S
A9518
Ampicillin sodium salt
C16H18N3NaO4S
A0166
Ampicillin sodium salt, powder, BioReagent, suitable for cell culture
C16H18N3NaO4S
A8351
Ampicillin sodium salt, BioXtra, suitable for cell culture
C16H18N3NaO4S
B3428 Anti-Bax antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500343
Anti-Bax antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA027878
Anti-BAX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502549
Anti-BAX antibody produced in rabbit, affinity isolated antibody
PRS3335
Anti-Bcl-2 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3337
Anti-Bcl-2 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300339
Anti-BCL2 (Ab-56) antibody produced in rabbit, affinity isolated antibody
SAB4300340
Anti-BCL2 (Ab-70) antibody produced in rabbit, affinity isolated antibody
AV01005
Anti-BCL2 antibody produced in rabbit, affinity isolated antibody
SAB2101603
Anti-NKX3-1 (ab2) antibody produced in rabbit, affinity isolated antibody
WH0000836M2
Monoclonal Anti-CASP3 antibody produced in mouse, clone 4D3, purified immunoglobulin, buffered aqueous solution
C5737 Monoclonal Anti-Caspase 3 antibody produced in mouse, clone 84803.111, purified immunoglobulin, lyophilized powder