EMAIL THIS PAGE TO A FRIEND
Allergy
Vitamin D deficiency induces Th2 skewing and eosinophilia in neonatal allergic airways disease.
PMID 24943330
Abstract
Associations between vitamin D status and childhood asthma are increasingly reported, but direct causation and mechanisms underlying an effect remain unknown. We investigated the effect of early-life vitamin D deficiency on the development of murine neonatal allergic airways disease (AAD). In utero and early-life vitamin D deficiency was achieved using a vitamin D-deficient diet for female mice during the third trimester of pregnancy and lactation. Offspring were weaned onto a vitamin D-deficient or vitamin D-replete diet, and exposure to intranasal house dust mite (HDM) or saline was commenced from day 3 of life for up to 6xa0weeks, when airway hyper-responsiveness (AHR), airway inflammation and remodelling were assessed. Neonatal mice that had in utero and early-life vitamin D deficiency had significantly increased pulmonary CD3(+) CD4(+) T1ST2(+) cells and reduced CD4(+) IL-10(+) cells. This effect was enhanced following HDM exposure. AHR in HDM-exposed mice was unaffected by vitamin D status. Introduction of vitamin D into the diet at weaning resulted in a significant reduction in serum IgE levels, reduced pulmonary eosinophilia and peri-bronchiolar collagen deposition. Peri-natal vitamin D deficiency alone has immunomodulatory effects including Th2 skewing and reduced IL-10-secreting T regulatory cells, exaggerated with additional allergen exposure. Vitamin D deficiency in early life does not affect AHR, but contributes to disease severity with worse eosinophilic inflammation and airway remodelling. Importantly, supplementation with vitamin D improves both of these pathological abnormalities.