EMAIL THIS PAGE TO A FRIEND
World journal of gastroenterology
Multispecies probiotic protects gut barrier function in experimental models.
PMID 24944474
Abstract
To investigate the effect of the probiotic combination Lactibiane Tolerance(®) (LT) on epithelial barrier function in vitro and in vivo. The effect of the multispecies probiotic LT was assessed on several models of epithelial barrier function both in vitro (in basal and inflammatory conditions) and in vivo [visceral hypersensitivity induced by chronic stress or by colonic perfusion of a fecal supernatant (FSN) from patients with irritable bowel syndrome (IBS)]. In vitro, we measured the permeability of confluent T84 cell monolayers incubated with or without LT by evaluating the paracellular flux of macromolecules, in basal conditions and after stimulation with lipopolysaccharide (LPS) or with conditioned medium of colonic biopsies from IBS patients (IBS-CM). In vivo, male C57/Bl6 mice received orally NaCl or LT for 15 d and were submitted to water avoidance stress (WAS) before evaluating visceral sensitivity by measuring the myoelectrical activity of the abdominal muscle and the paracellular permeability with (51)Cr-EDTA. Permeability and sensitivity were also measured after colonic instillation of FSN. Tight-junctions were assessed by immunoblotting and TLR-4 expression was evaluated by immunohistochemistry Incubation of T84 cell monolayers with LT in basal conditions had no significant effect on permeability (P > 0.05 vs culture medium). By contrast, addition of LT bacterial bodies (LT) completely prevented the LPS-induced increase in paracellular permeability (P < 0.01 vs LPS 10 ng/mL (LPS 10); P < 0.01 vs LPS 100 ng/mL (LPS 100), P > 0.05 vs culture medium). The effect was dose dependent as addition of 10(9) LT bacterial bodies induced a stronger decrease in absorbance than 10(6) LT (10(9) LT + LPS 10: -20.1% ± 13.4, P < 0.01 vs LPS 10; 10(6) LT + LPS 10: -11.6% ± 6.2, P < 0.01 vs LPS 10; 10(9) LT + LPS 100: -14.4% ± 5.5, P < 0.01 vs LPS 100; 10(6) LT + LPS 100: -11.6% ± 7.3, P < 0.05 vs LPS 100). Moreover, the increase in paracellular permeability induced by culturing T84 cells with conditioned medium of colonic biopsies from IBS patients (IBS-CM) was completely inhibited in the presence of 10(9) LT (P < 0.01 vs IBS-CM). LT also significantly prevented the epithelial disruption induced by intracolonic infusion of fecal supernatant from IBS patients (P < 0.01 vs IBS FSN) or water avoidance stress P < 0.01 vs WAS) in C57/Bl6 mice and increased the expression of occludin in vitro and in vivo, as assessed by immnunoblotting. The WAS-induced effect on visceral sensitivity was prevented by LT treatment since values obtained for all steps of colorectal distension were significantly (P < 0.01) different from the WAS group. Finally, LT down-regulated the response mediated through TLR-4 in vitro (decrease in tumor necrosis factor α secretion in response to LPS: -65.8% for 10(9) LT and -52.5% for 10(6) LT, P < 0.01 vs LPS) and in vivo (inhibition of WAS induced an increase in TLR-4 expression in the LT treated mice colon, P < 0.01 vs WAS). The probiotic LT mix prevented the disruption to the epithelial barrier induced by LPS, stress or colonic soluble factors from IBS patients and prevented visceral hypersensitivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7617
Anti-Calnexin antibody, Mouse monoclonal, clone TO-5, purified from hybridoma cell culture
SAB2501291
Anti-CANX antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA009433
Anti-CANX antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA009696
Anti-CANX antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB5200084
Anti-CANX antibody produced in rabbit, whole antiserum
SAB5200097 Anti-CANX antibody produced in rabbit, affinity isolated antibody
SAB1406198
Anti-OCLN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2501634
Anti-TJP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA001636
Anti-TJP1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102434
Anti-TJP1 antibody produced in rabbit, affinity isolated antibody
HPA001637
Anti-TJP1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB5300526
Monoclonal Anti-CANX antibody produced in mouse, clone 3H4A7, ascites fluid
WH0004950M1
Monoclonal Anti-OCLN antibody produced in mouse, clone 1G7, purified immunoglobulin, buffered aqueous solution
AMAB90893
Monoclonal Anti-OCLN antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1608, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90890
Monoclonal Anti-OCLN antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1567, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90889
Monoclonal Anti-OCLN antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1555, purified immunoglobulin, buffered aqueous glycerol solution