EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
JNK inhibitor SP600125 protects against lipopolysaccharide-induced acute lung injury via upregulation of claudin-4.
PMID 24944614
Abstract
Although