EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical and Biomedical Analysis
Molecularly imprinted polymer for specific extraction of hypericin from Hypericum perforatum L. herbal extract.
PMID 24946147
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31573
5α-Androstan-17β-ol-3-one, VETRANAL, analytical standard
C19H30O2
71251
Acetic acid, analytical standard
C2H4O2
45983
Acetonitrile, analytical standard
C2H3N
PHR1551
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
00712
Acetonitrile solution, ~20% in H2O, for protein sequence analysis
C2H3N
00213
Benzoyl peroxide blend with dicyclohexyl phthalate, technical, ~50% (T)
C14H10O4
B5907
Benzoyl peroxide blend with dicyclohexyl phthalate, suitable for use as a catalyst for electron microscopy. Modified to render it safe in transit.
C14H10O4
90577
Bifido Selective Supplement B, for microbiology
C2H4O2
1005706
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
C2H4O2
632651
Luperox® A70S, Benzoyl peroxide, 70%, remainder water
C14H10O4
517909
Luperox® A75, Benzoyl peroxide, 75%, remainder water
C14H10O4
513474
Luperox® A75FP, Benzoyl peroxide, 75% remainder water, contains 25 wt. % water as stabilizer, 75%
C14H10O4
179981
Luperox® A98, Benzoyl peroxide, reagent grade, ≥98%
C14H10O4
513431
Luperox® AFR40, Benzoyl peroxide solution, 40 wt. % in dibutyl phthalate
C14H10O4
513466
Luperox® ATC50, Benzoyl peroxide, ~50 wt. % in tricresyl phosphate
C14H10O4
82762
Methanol, analytical standard
CH4O
PHR1372
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
CH4O
1424109
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
CH4O
D145750
N,N-Dimethylaniline, 99%
C8H11N
407275
N,N-Dimethylaniline, purified by redistillation, ≥99.5%
C8H11N
515124
N,N-Dimethylaniline, ReagentPlus®, 99%
C8H11N
PHR1320
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
1601340
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
C2H3N