EMAIL THIS PAGE TO A FRIEND
Pathology, research and practice
Effect of BRCA1 immunohistochemical localizations on prognosis of patients with sporadic breast carcinomas.
PMID 24947414
Abstract
Our purpose was to investigate the expression pattern of BRCA1 protein in sporadic breast carcinomas, as well as the clinicopathological and prognostic value of its subcellular localizations. Immunohistochemistry was performed on paraffin embedded tissue specimens from 111 sporadic, invasive breast carcinomas to detect the expression of the proteins BRCA1, ER, PR, erbB2, p53 and Ki67. BRCA1 protein was detected in the nuclei and the cytoplasm of the tumor cells. Nuclear BRCA1 immunoreactivity showed no relation with the classic clinicopathological markers and the expression of cerbB2, p53 and Ki67. Reduced expression of nuclear BRCA1 protein was found to exert an independent favorable impact on both the overall and relapse-free (RF) survival of the patients (p=0.019 and p=0.043, respectively). Cytoplasmic BRCA1 was associated with none of the classic histomorphological indices, except from the lymph node metastasis, with which its relation was found to be inverse (p=0.05), prolonging the RF survival of the patients (p=0.05). Our findings suggest that BRCA1 protein depicts different prognostic significance, according to its subcellular distribution. Nuclear detection of the protein was associated with a worse prognosis, while the cytoplasmic one was related to fewer recurrences as a result of fewer lymph node metastases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA000451
Anti-MKI67 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001164
Anti-MKI67 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306667
ANTI-P53 (S15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306713
ANTI-P53 (S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
P4235 Anti-p53 antibody produced in sheep, fractionated antiserum, buffered aqueous solution
P6874
Anti-p53 antibody, Mouse monoclonal, clone DO-1, purified from hybridoma cell culture
P7874
Anti-p53 antibody, Mouse monoclonal, clone DO-2, purified from hybridoma cell culture
SAB4300348
Anti-PGR (Ab-190) antibody produced in rabbit, affinity isolated antibody
HPA004751
Anti-PGR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA008428
Anti-PGR antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA017176
Anti-PGR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW22679 Anti-Progesterone Receptor antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4502184
Anti-Progesterone Receptor antibody produced in rabbit, affinity isolated antibody
SAB4502185
Anti-Progesterone Receptor antibody produced in rabbit, affinity isolated antibody
SAB4502186
Anti-Progesterone Receptor antibody produced in rabbit, affinity isolated antibody
HPA053794
Anti-TMEM37 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300365
Anti-TP53 (Ab-18) antibody produced in rabbit, affinity isolated antibody
SAB4300369
Anti-TP53 (Ab-315) antibody produced in rabbit, affinity isolated antibody
SAB4300366
Anti-TP53 (Ab-33) antibody produced in rabbit, affinity isolated antibody
SAB4300367
Anti-TP53 (Ab-37) antibody produced in rabbit, affinity isolated antibody
SAB4300368
Anti-TP53 (Ab-46) antibody produced in rabbit, affinity isolated antibody
SAB4300362
Anti-TP53 (Ab-6) antibody produced in rabbit, affinity isolated antibody
SAB4300363
Anti-TP53 (Ab-9) antibody produced in rabbit, affinity isolated antibody
SAB1406544
Anti-TP53 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV02055
Anti-TP53 antibody produced in rabbit, IgG fraction of antiserum
SAB2106418
Anti-TRP53 antibody produced in rabbit, affinity isolated antibody
P5813
Monoclonal Anti-p53 antibody produced in mouse, clone BP53-12, ascites fluid
P6749
Monoclonal Anti-p53 antibody produced in mouse, clone PAb1801, purified from hybridoma cell culture
P8999
Monoclonal Anti-p53 antibody produced in mouse, clone DO-7, purified from hybridoma cell culture
AMAB90956
Monoclonal Anti-p53 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2199, purified immunoglobulin, buffered aqueous glycerol solution