EMAIL THIS PAGE TO A FRIEND
Arteriosclerosis, thrombosis, and vascular biology
Factor XI regulates pathological thrombus formation on acutely ruptured atherosclerotic plaques.
PMID 24947525
Abstract
Coagulation factor XI is proposed as therapeutic target for anticoagulation. However, it is still unclear whether the antithrombotic properties of factor XI inhibitors influence atherosclerotic disease and atherothrombosis. Our aim is to investigate whether factor XI antisense oligonucleotides could prevent thrombus formation on acutely ruptured atherosclerotic plaques. Atherosclerotic plaques in the carotid arteries of Apoe(-/-) mice were acutely ruptured using ultrasound. The subsequent thrombus formation was visualized and quantified by intravital microscopy and immunohistochemistry. Mice were pretreated with either factor XI antisense or nonsense oligonucleotides (50 mg/kg) to lower factor XI plasma levels. A tail bleeding assay was used to determine the safety. On plaque rupture, initial platelet adhesion and platelet plug formation were not impaired in animals treated with factor XI antisense oligonucleotides. However, the ensuing thrombus formation and fibrin deposition were significantly lower after 5 to 10 minutes (P<0.05) in factor XI antisense oligonucleotide-treated animals without inducing a bleeding tendency. Furthermore, thrombi from antisense-treated animals were less stable than thrombi from placebo-treated animals. Moreover, macrophage infiltration and collagen deposition were lower in the carotid arteries of factor XI antisense-treated animals. No neutrophils were present. Factor XI antisense oligonucleotides safely prevent thrombus formation on acutely ruptured atherosclerotic plaques in mice. Furthermore, perturbed carotid arteries from factor XI antisense-treated animals show a less severe inflammatory response.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9879 Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
C7806 Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
C1188 Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
C8919 Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
C9791 Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
C9301 Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
C0543 Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder (from 0.2 μm-filtered solution), BioReagent, suitable for cell culture
C7521
Collagen from human placenta, Bornstein and Traub Type IV, powder
C4407 Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
C5533
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
C7774 Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
C3657 Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
C8374
Collagen from human placenta, Bornstein and Traub Type IV, powder
H4417 Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, ≥95% (SDS-PAGE)
C5608 Collagen from rabbit skin, Bornstein and Traub Type I, powder
C7661 Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
27666 Collagen from rat tail, for cell biology, soluble
C5483
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
C7624
Collagen human, Bornstein and Traub Type I, recombinant, expressed in Nicotiana tabacum (tobacco)
C7999
Collagen human, Bornstein and Traub Type I, recombinant, expressed in Nicotiana tabacum (tobacco)
C8993 Collagen ovine, aqueous solution
C6745 Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
C3867 Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
F5386 Fibrin from human plasma, insoluble powder