EMAIL THIS PAGE TO A FRIEND
Pharmacological reports : PR
Investigation into the role of Cu/Zn-SOD delivery system on its antioxidant and antiinflammatory activity in rat model of peritonitis.
PMID 24948070
Abstract
The current study evaluated the role of delivery system (solution, conventional liposomes and PEG-ylated liposomes) on superoxide dismutase (SOD) antioxidant and antiinflammatory properties in a rat model of lipopolysaccharide (LPS)-induced peritonitis. Fifty male albino rats (Wistar-Bratislava) were divided into five groups (n=10). Control group received saline and the other four groups received intraperitoneal injections of LPS (5mg/kg). Among the LPS-injected groups, one was LPS control group and the other three groups received the endotoxin injection 30min after receiving the same dose of SOD (500U/kg, ip) in different delivery systems: saline solution (SOD-S), conventional liposomes (SOD-L) or PEG-ylated liposomes (SOD-PL). The animals were euthanized 6h after LPS injection, blood samples were collected and acute phase response (total and differential leukocytes count; tumor necrosis factor α), antioxidants (total antioxidants; reduced glutathione), oxidative stress (total oxidants; lipid peroxidation) and nitrosative stress (nitric oxide metabolites; nitrotyrosine) were evaluated. Intraperitoneal administration of LPS to rats induced a marked inflammatory and oxidative response in plasma. On the other hand, all SOD formulations had protective effect against endotoxin-induced inflammation and oxidative/nitrosative stress, but PEG-ylated liposomes had the most significant activity. Thus, SOD-PL administration significantly reduced the effects of LPS on bone marrow acute phase response, the oxidative status and production of nitric oxide metabolites, while increasing the markers of antioxidant response in a significant manner. SOD supplementation interferes both with inflammatory and oxidative pathways involved in LPS-induced acute inflammation, PEG-ylated liposomal formulation being of choice among the tested delivery systems.