EMAIL THIS PAGE TO A FRIEND
Analytical and Bioanalytical Chemistry
Identification and monitoring of thiabendazole transformation products in water during Fenton degradation by LC-QTOF-MS.
PMID 24948093
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
194123
Benzimidazole, 98%
C7H6N2
CDS006757
Thiazole-4-carboxamide, AldrichCPR
C4H4N2OS