EMAIL THIS PAGE TO A FRIEND
AMB Express
Cloning, expression and characterization of a versatile Baeyer-Villiger monooxygenase from Dietzia sp. D5.
PMID 24949258
Abstract
A novel BVMO encoding gene was identified from a draft genome sequence of a newly isolated strain of Dietzia. Analysis of the protein sequence revealed that it belongs to a group of BVMOs whose most characterized member is cyclopentadecanone monooxygenase (CPDMO). The gene was PCR amplified, cloned and successfully expressed in E. coli. The expressed recombinant enzyme was purified using metal affinity chromatography. Characterization of the purified enzyme revealed that it has a broad substrate scope and oxidized different compounds including substituted and unsubstituted alicyclic, bicyclic-, aliphatic-ketones, ketones with an aromatic moiety, and sulfides. The highest activities were measured for 2- and 3-methylcyclohexanone, phenylacetone, bicyclo-[3.2.0]-hept-2-en-6-one and menthone. The enzyme was optimally active at pHxa07.5 and 35°C, a temperature at which its half-life was about 20xa0hours. The stability studies have shown that this enzyme is more stable than all other reported BVMOs except the phenylacetone monooxygenase from the thermophilic organism Thermobifida fusca.