EMAIL THIS PAGE TO A FRIEND
Environmental Research
Persistent organic pollutant levels in human visceral and subcutaneous adipose tissue in obese individuals--depot differences and dysmetabolism implications.
PMID 24949816