EMAIL THIS PAGE TO A FRIEND
Analytical and Bioanalytical Chemistry
Continuous vs. segmented second-dimension system gradients for comprehensive two-dimensional liquid chromatography of sugarcane (Saccharum spp.).
PMID 24951131
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5893
(±)-Naringenin, ≥95%
C15H12O5
52186
(±)-Naringenin, analytical standard
C15H12O5
C0625
Caffeic acid, ≥98.0% (HPLC)
C9H8O4
60018
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
C9H8O4
60020
Caffeic acid, purum, ≥95.0% (HPLC)
C9H8O4
C3878
Chlorogenic acid, ≥95% (titration)
C16H18O9
00500590
Chlorogenic acid, primary pharmaceutical reference standard
C16H18O9
Y0000569
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
C16H18O9
72609
Coumarin, certified reference material, TraceCERT®
C9H6O2
C4261
Coumarin, ≥99% (HPLC)
C9H6O2
W526509
Coumarin, ≥98%
C9H6O2
01260595
Coumarin, primary pharmaceutical reference standard
C9H6O2
Y0000438
Coumarin, European Pharmacopoeia (EP) Reference Standard
C9H6O2
G7384
Gallic acid, 97.5-102.5% (titration)
C7H6O5
91215
Gallic acid, certified reference material, TraceCERT®
C7H6O5
77634
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
C8H8O3
W530098
Naringenin, natural (US), 98%
C15H12O5
30304
Vanillin, TraceCERT®, certified reference material
C8H8O3
94752
Vanillin, tested according to Ph.Eur.
C8H8O3
PHR1245
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
C8H8O3
V1104
Vanillin, ReagentPlus®, 99%
C8H8O3
W310700
Vanillin, ≥97%, FCC, FG
C8H8O3
W310727
Vanillin, natural, ≥97%, FCC, FG
C8H8O3
V0050000
Vanillin, European Pharmacopoeia (EP) Reference Standard
C8H8O3
PHR1091
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
C8H8O3
1711009
Vanillin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
C8H8O3