EMAIL THIS PAGE TO A FRIEND
AJR. American journal of roentgenology
Potential role of 18F-FDG PET/CT in patients with fungal infections.
PMID 24951213
Abstract
Combined anatomic and functional imaging with (18)F-FDG PET/CT is slowly gaining foothold in the management of various infective pathologic abnormalities. However, limited literature is available regarding the role of FDG PET/CT in patients with fungal infections. Here, we briefly review the available literature and highlight the potential role that FDG PET/CT can play in the diagnosis and management of fungal infections.