EMAIL THIS PAGE TO A FRIEND
Journal of Pharmacology and Experimental Therapeutics
Astragaloside IV ameliorates renal fibrosis via the inhibition of mitogen-activated protein kinases and antiapoptosis in vivo and in vitro.
PMID 24951279