EMAIL THIS PAGE TO A FRIEND
The Journal of Antimicrobial Chemotherapy
Mutations in the chikungunya virus non-structural proteins cause resistance to favipiravir (T-705), a broad-spectrum antiviral.
PMID 24951535
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9415
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
C62H86N12O16
A4262
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
C62H86N12O16
746460
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
ClLi
793620
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
ClLi
L4408
Lithium chloride, BioXtra, ≥99.0% (titration)
ClLi
62476
Lithium chloride, BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)
ClLi
L9650
Lithium chloride, for molecular biology, ≥99%
ClLi
429457
Lithium chloride, anhydrous, beads, −10 mesh, 99.998% trace metals basis
ClLi
449881
Lithium chloride, anhydrous, beads, −10 mesh, ≥99.9% trace metals basis
ClLi
203637
Lithium chloride, powder, ≥99.99% trace metals basis
ClLi
HT-LICLSLUG
Lithium chloride, anhydrous, chunks, 99.99% trace metals basis
ClLi
751898
Lithium chloride, anhydrous, chunks, 99.99% trace metals basis
ClLi
L7026
Lithium chloride solution, 8 M, for molecular biology, ≥99%
ClLi
694576
Lithium chloride solution, 1 M in 1,3-dimethyl-2-imidazolidinone
ClLi
62483
Lithium chloride solution, 1 M in glacial acetic acid
ClLi
62481
Lithium chloride solution, 1 M in ethanol
ClLi
667544
Lithium chloride solution, 0.5 M in anhydrous tetrahydrofuran
ClLi
LCL30
Lithium chloride solution, 2 M in ethanol
ClLi
601489
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Cl7Li
P9625
Phenazine methosulfate, ≥90% (UV)
C14H14N2O4S
1613655
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
CHNaO3
71631
Sodium bicarbonate, tested according to Ph.Eur.
CHNaO3