EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Spatial and temporal distribution of chiral pesticides in Calanus spp. from three Arctic fjords.
PMID 24951967
Abstract
Concentration and enantiomeric fractions (EFs) of chiral chlorinated pesticides (α-hexachlorocyclohexane (α-HCH), trans-, cis- and oxychlordane) were determined in Arctic zooplankton, mainly Calanus spp. collected in the period 2007-11 from Svalbard fjords and open pack-ice. The temporal and spatial enantiomer distribution varied considerably for all species and chiral pesticides investigated. An overall enantiomeric excess of (+)-oxychlordane (EF 0.53-0.86) were observed. Cis-chlordane was close to racemic (EF 0.46-0.55), while EF for trans-chlordane varied between 0.29 and 0.55, and between 0.38 and 0.59 for α-HCH. The biodegradation potential for trans-chlordane was higher compared to cis-chlordane. The comprehensive statistical evaluation of the data set revealed that the EF distribution of α-HCH was affected by ice cover to a higher extent compared to cis-chlordane. Potential impact from benthic processes on EFs in zooplankton is an interesting feature and should be further investigated. Enantiomeric selective analyses may be a suitable tool for investigations of climate change related influences on Arctic ecosystems.