Histone deacetylase 2 and N-Myc reduce p53 protein phosphorylation at serine 46 by repressing gene transcription of tumor protein 53-induced nuclear protein 1.
PMID 24952595
Myc oncoproteins and histone deacetylases (HDACs) exert oncogenic effects by modulating gene transcription. Paradoxically, N-Myc induces p53 gene expression. Tumor protein 53-induced nuclear protein 1 (TP53INP1) phosphorylates p53 protein at serine 46, leading to enhanced p53 activity, transcriptional activation of p53 target genes and programmed cell death. Here we aimed to identify the mechanism through which N-Myc overexpressing p53 wild-type neuroblastoma cells acquired resistance to apoptosis. TP53INP1 was found to be one of the genes most significantly repressed by HDAC2 and N-Myc according to Affymetrix microarray gene expression datasets. HDAC2 and N-Myc reduced TP53INP1 gene expression by direct binding to the TP53INP1 gene promoter, leading to transcriptional repression of TP53INP1, p53 protein de-phosphorylation at serine 46, neuroblastoma cell proliferation and survival. Moreover, low levels of TP53INP1 expression in human neuroblastoma tissues correlated with high levels of N-Myc expression and poor patient outcome, and the BET bromodomain inhibitors JQ1 and I-BET151 reduced N-Myc expression and reactivated TP53INP1 expression in neuroblastoma cells. These findings identify TP53INP1 repression as an important co-factor for N-Myc oncogenesis, and provide further evidence for the potential application of BET bromodomain inhibitors in the therapy of N-Myc-induced neuroblastoma.