EMAIL THIS PAGE TO A FRIEND
Oncotarget
XIAP inhibitors induce differentiation and impair clonogenic capacity of acute myeloid leukemia stem cells.
PMID 24952669
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3768
Dequalinium chloride hydrate, ≥95% (titration)
C30H40Cl2N4 · xH2O