EMAIL THIS PAGE TO A FRIEND
Journal of Ethnopharmacology
Effects of San-Huang-Xie-Xin-tang, a traditional Chinese prescription for clearing away heat and toxin, on the pacemaker activities of interstitial cells of Cajal from the murine small intestine.
PMID 24953035