EMAIL THIS PAGE TO A FRIEND
Molecular Genetics and Metabolism
A CLN8 nonsense mutation in the whole genome sequence of a mixed breed dog with neuronal ceroid lipofuscinosis and Australian Shepherd ancestry.
PMID 24953404