EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
Quantification of levornidazole and its metabolites in human plasma and urine by ultra-performance liquid chromatography-mass spectrometry.
PMID 24953495