EMAIL THIS PAGE TO A FRIEND
Arch. Pharm. (Weinheim)
In vitro and in silico studies to explore structural features of flavonoids for aldehyde oxidase inhibition.
PMID 24956178
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5893
(±)-Naringenin, ≥95%
C15H12O5
52186
(±)-Naringenin, analytical standard
C15H12O5
D7802
Daidzein, ≥98%, synthetic
C15H10O4
16587
Daidzein, analytical standard
C15H10O4
92136
Genistein, analytical standard
C15H10O5
H4125
Hesperetin, analytical standard
C16H14O6
W431300
Hesperetin, ≥95%
C16H14O6
H5254
Hesperidin, ≥80%
C28H34O15
50162
Hesperidin, analytical standard
C28H34O15
Y0001203
Hesperidin, European Pharmacopoeia (EP) Reference Standard
C28H34O15
04650590
Hesperidin, primary pharmaceutical reference standard
C28H34O15
M9429
Menadione, meets USP testing specifications
C11H8O2
1381006
Menadione, United States Pharmacopeia (USP) Reference Standard
C11H8O2
M0300000
Menadione, European Pharmacopoeia (EP) Reference Standard
C11H8O2
PHR1411
Menadione, Pharmaceutical Secondary Standard; Certified Reference Material
C11H8O2
47775
Menadione (K3), analytical standard
C11H8O2
72576
Myricetin, analytical standard
C15H10O8
M6760
Myricetin, ≥96.0%, crystalline
C15H10O8
70050
Myricetin, ≥96.0% (HPLC)
C15H10O8
W530098
Naringenin, natural (US), 98%
C15H12O5
N1376
Naringin, ≥90%, from citrus fruit
C27H32O14
71162
Naringin, ≥95% (HPLC)
C27H32O14
91842
Naringin, analytical standard
C27H32O14
Y0001378
Naringin, European Pharmacopoeia (EP) Reference Standard
C27H32O14
60032
Potassium standard for AAS, ready-to-use, traceable to BAM, in H2O
ClK
49513
Vitexin, analytical standard
C21H20O10
00840595
Vitexin, primary pharmaceutical reference standard
C21H20O10