EMAIL THIS PAGE TO A FRIEND
Journal of Translational Medicine
Variability in CRP, regulatory T cells and effector T cells over time in gynaecological cancer patients: a study of potential oscillatory behaviour and correlations.
PMID 24957270