EMAIL THIS PAGE TO A FRIEND
Psychopharmacology Series
Stress-induced activation of ventral tegmental mu-opioid receptors reduces accumbens dopamine tone by enhancing dopamine transmission in the medial pre-frontal cortex.
PMID 24958228