EMAIL THIS PAGE TO A FRIEND
Journal of psychopharmacology (Oxford, England)
Involvement of neuronal nitric oxide synthase in desensitisation of µ-opioid receptors in the rat locus coeruleus.
PMID 24961237
Abstract
Nitric oxide (NO) has been recently shown to enhance µ-opioid receptor (MOR) desensitisation in locus coeruleus (LC) neurons. The aim of this study was to evaluate by single-unit extracellular recordings in rat brain slices whether the neuronal NO synthase is involved in MOR desensitisation in LC neurons. As expected, a high concentration of the opioid agonist Met(5)-enkephalin (ME; 10 µM, 10 min) strongly desensitised the inhibition induced by a test application of ME (0.8 µM, 1 min), whereas lower ME concentrations (1 and 3 µM) only weakly desensitised it. The neuronal NO synthase inhibitors 7-nitroindazole (10-100 µM), S-methyl-L-thiocitrulline (0.01-10 µM) and N(ω)-propyl-L-arginine (1-10 µM) attenuated ME (10 µM)-induced opioid desensitisation, although the endothelial NO synthase inhibitor N(5)-(1-iminoethyl)-L-ornithine (3-30 µM) failed to change it. The NO donor sodium nitroprusside (1 mM), but not its inactive analog potassium ferricyanide (1 mM), enhanced the ME (3 µM)-induced desensitisation and prevented the effect of S-methyl-L-thiocitrulline (10 µM). Sodium nitroprusside (1 mM) failed to change the desensitisation of α2-adrenoceptors by noradrenaline (100 µM, 10 min). These results suggest the contribution of NO and a neuronal type of NO synthase in homologous MOR desensitisation in rat LC neurons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7778
7-Nitroindazole, ≥98%
C7H5N3O2